missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Chloramphenicol Selective Supplement

Product Code. 11464159
Change view
Click to view available options
Yield:
Each vial for 2L medium
Each vial for 500mL medium
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Yield unitSize
11464159 Each vial for 500mL medium Pack of 10
12771860 Each vial for 2L medium Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464159 Supplier Thermo Scientific™ Supplier No. SR0078E
  • ONLINE EXCLUSIVE PRICE
    42.05 €
    38.31 € / Pack of 10
    Save 3.74 €
    8% Off
View all promotions
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Oxoid Chloramphenicol Selective Supplement assists in the isolation of yeast and molds associated with spoilage.

For the isolation of yeast and molds associated with spoilage, add Oxoid Chloramphenicol Selective Supplement to DRBC Agar Base, Part No. CM0727B; for the enumeration of yeast and molds in foods, add to Rose Bengal Chloramphenicol Agar Base, Part No. CM0549B.

For the isolation of yeast and molds associated with spoilage, add to DRBC Agar Base, Part No. CM0727B; for the enumeration of yeast and molds in foods, add to Rose Bengal Chloramphenicol Agar Base, Part No. CM0549B.
TRUSTED_SUSTAINABILITY

Specifications

Description Chloramphenicol Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.