missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ CO2Gen™ Compact Sachet

Product Code. 10565953
Change view
Click to view available options
Quantity:
20/Pk.
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10565953 20/Pk. Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10565953 Supplier Thermo Scientific™ Supplier No. CD0020C
  • ONLINE EXCLUSIVE PRICE
    73.15 €
    71.35 € / Pack of 20
    Save 1.80 €
    2% Off
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Oxoid CO2 Gen Compact Sachet is for use in Compact Plastic Pouch.

Oxoid CO2 Gen Compact Sachet is for use in Compact Plastic Pouch.

  • No water or catalyst required
  • No hydrogen gas produced
  • For use with Compact Plastic Pouch, Part No. AG0020C and AnaeroGen Compact Sealing Clip, Part No. AN0005C
TRUSTED_SUSTAINABILITY

Specifications

Type CO2 Generating Compact Sachet
Format Compact Gas Generation Sachet
Product Line Thermo Scientific Oxoid
Quantity 20/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.