missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Cycloheximide Solution (0.1%)

Product Code. 10661185
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10661185 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10661185 Supplier Thermo Scientific™ Supplier No. SR0222C
  • ONLINE EXCLUSIVE PRICE
    39.46 €
    35.23 € / 100mL
    Save 4.23 €
    10% Off
View all promotions
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Oxoid Cycloheximide Solution (0.1%) (100mg/vial) is used for the suppression of fungi.

Oxoid Cycloheximide Solution (0.1%) (100mg/vial) is used for the suppression of fungi.

Add to Bacillus Cereus Selective Agar Base, Part No. CM0617B; Raka-Ray Agar Base, Part No. CM0777B; and Universal Beer Agar, Part No. CM0651B
TRUSTED_SUSTAINABILITY

Specifications

Description Cycloheximide Solution (0.1%)
Form Liquid
Product Type Solution
Quantity 100 mL
Yield Yield will be dependent upon formulation; follow the guidelines for your specific product.
compliance-icons
Product Identifier
  • CYCLOHEXIMIDE SOLUTION
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
Supplemental information
  • MIXTURE LIST-Contain: Cycloheximide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.