missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Diagnostic Sensitivity Test Agar (Dehydrated)

Product Code. 11923462
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11923462 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11923462 Supplier Thermo Scientific™ Supplier No. CM0261B
  • ONLINE EXCLUSIVE PRICE
    167.84 €
    156.65 € / 500g
    Save 11.19 €
    6% Off
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform effortless Antimicrobial Susceptibility Testing (AST) of fastidious pathogens with this medium.

Perform effortless Antimicrobial Susceptibility Testing (AST) of fastidious pathogens with Thermo Scientific™ Oxoid™ Diagnostic Sensitivity Test Agar (Dehydrated). Diagnostic Sensitivity Test Agar was developed as a dual purpose medium which would satisfy both diagnostic and susceptibility requirements.

Diagnostic Sensitivity Test Agar is recommended for diagnostic as well as testing of susceptibility of organisms to antibiotics. The diagnostic role is supported by the nutritional amino-acid base with glucose to encourage early growth. The inclusion of the buffers (disodium phosphate and sodium acetate) help prevent excessive movements of pH values which could result from utilization of glucose or amino acids. Such pH movements would interfere with haemolytic reactions1 and the MIC values of pH-susceptible antimicrobials2.

The agar used in the formulation has been specially processed to allow unimpeded diffusion of antimicrobials from discs3.

Diagnostic Sensitivity Test Agar is now primarily used for susceptibility tests and its role in diagnostic microbiology, i.e. the primary isolation of organisms from clinical samples, has diminished.

An essential requirement for satisfactory antimicrobial susceptibility media is that the reactive levels of thymidine and thymine must be sufficiently reduced to avoid antagonism of trimethoprim and sulphonamides4. Diagnostic Sensitivity Test Agar meets this requirement and in the presence of lysed horse blood (or defibrinated horse blood if the plates are stored long enough to allow some lysis of the erythrocytes) the level of thymidine will be further reduced. This is caused by the action of the enzyme thymidine phosphorylase which is released from lysed horse erythrocytes5. Thymidine is an essential growth factor for thymidine-dependent organisms and they will not grow in its absence or they will grow poorly in media containing reduced levels6.

TRUSTED_SUSTAINABILITY

Specifications

Description Diagnostic Sensitivity Test Agar (DSTA)
Form Powder
Product Type Agar
Quantity 500 g
Yield For 12.5L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.