missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DNASE Agar (Dehydrated)

Product Code. 10566323
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10566323 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10566323 Supplier Thermo Scientific™ Supplier No. CM0321B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Detect the microbial deoxyribonuclease enzymes, particularly from staphylococci with this differential medium.

Detect microbial deoxyribonuclease enzymes, particularly from staphylococci, with Thermo Scientific™ Oxoid™ DNASE Agar (Dehydrated). A DNase (deoxyribonuclease) is any enzyme that catalyzes the cleavage of phosphodiester linkages of DNA leading to its degradation. The organisms producing DNase enzymes in sufficient quantities to hydrolyze DNA will show clear zones around the colonies. The medium is mainly used in the identification of staphylococci but may also be used for the detection of DNase activity in other organisms.

Weckman & Catlin1 suggested that DNase activity could be used to identify pathogenic staphylococci after they had established a close correlation with coagulase production. Jeffries et al.2 incorporated DNA in the agar medium to provide a simple method of detecting DNase activity.

  • Easily differentiates Staphylococcus aureus from Staphylococcus epidermidis and other Staphylococcus species due to the abundance of DNase production in Staphylococcus aureus. 2,3,4
  • Helps in the differentiation and identification of non-pigmented Serratia marcescens (positive reaction) from Klebsiella-Enterobacter (negative reaction)5.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description DNase Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 12.8L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.