Learn More
Thermo Scientific™ Oxoid™ Florfenicol Antimicrobial Susceptibility Disks, 30 μg
Product Code. 10361253
Change view
Click to view available options
Concentration:
30 μg
Unit Size:
Pack of 250
Product Code. Concentration Unit Size
10361253 30 μg Pack of 250
1 options
Product Code. 10361253 Supplier Thermo Scientific™ Supplier No. CT1754B
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Determine the susceptibility of microorganisms using manual AST methods and a full range of high-quality, easy-to-use antimicrobial susceptibility testing disks.
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Florfenicol Antimicrobial susceptibility Disks.
Ease-of-use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
Disk diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Florfenicol
Certifications/Compliance Complies with CLSI guidelines
Concentration 30 μg
Description Oxoid Florfenicol Antimicrobial Susceptibility Disks
Antibiotic Code FFC
Product Type Antimicrobial Susceptibility Discs
Test Type Antimicrobial Susceptibility
No. per Pack 5 Cartridges/Pk.
Packaging Type 50 Disks/Cartridge
Quantity 50 Discs/Cartridge