missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Gardnerella Vaginalis Selective Supplement
Description
Oxoid Gardnerella Vaginalis Selective Supplement is used for the isolation of Gardnerella vaginalis.
- Add to to Columbia Blood Agar Base, Part No. CM0331B
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | Gardnerella Vaginalis Selective Supplement |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |
| Species | Gardnerella vaginalis |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?