missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Heart Infusion Broth (Dehydrated)

Product Code. 10371613
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10371613

Brand: Thermo Scientific™ CM1032B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cultivate streptococci, pneumococci, meningococci and other fastidious organisms with this nutritious medium.

Cultivate streptococci, pneumococci, meningococci and other fastidious organisms with Thermo Scientific™ Oxoid™ Heart Infusion Broth (Dehydrated). The medium is highly nutritious and well buffered to support the growth of wide variety of organisms.

Heart Infusion Broth is developed to give the same performance characteristics as Brain Heart Infusion (BHI) Broth. However, as bovine brain is a specified risk material, the exclusion of it from the Heart Infusion Broth means that it carries a lower regulatory burden.

Cultivate a wide variety of organisms with Heart Infusion Broth.

  • Developed to give equivalent performance to Brain Heart Infusion Broth while significantly reducing the regulatory burden due to the exclusion of calf brain infusion.
  • Addition of anti-microbial agents such as cycloheximide and chloramphenicol make this medium suitable for the cultivation of yeasts and molds.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Thermo Scientific Oxoid Heart Infusion Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 13.5L medium

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.