missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Kligler Iron Agar (Dehydrated)

Product Code. 10631675
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
Product Code. Quantity unitSize
10631675 500 g 500g
1 options
This item is not returnable. View return policy

Product Code. 10631675

Brand: Thermo Scientific™ CM0033B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Identify Enterobacteriaceae on the basis of double sugar fermentation and hydrogen sulphide production with this medium.

Identify Enterobacteriaceae on the basis of double sugar fermentation and hydrogen sulphide production using Thermo Scientific™ Oxoid™ Kligler Iron Agar (Dehydrated). Klingler Iron Agar combines the features of Kligler's Lead Acetate medium1,2,3 and Russell's Double Sugar Agar4. The medium is recommended for the identification of colonies picked off from plating media such as MacConkey Agar, Bismuth Sulphite Agar, or Desoxycholate Citrate Agar, etc.

Differentiate Enterobacteriaceae on the basis of double sugar fermentation and hydrogen sulphide production with Klinger Iron Agar.

Non lactose fermenters initially produce a yellow slant and butt as a result of glucose fermentation. Once the glucose is depleted, the reactions reverts to red slants (alkaline) due to oxidation of acids. Lactose fermenting organisms produce yellow slants and butts. There is no reversion to red in the slant because enough acid is produced to maintain an acid pH under aerobic conditions. Non-fermenters produce red slants and butts. H 2 S production results in a blackening of the medium, either throughout the butt or in a ring formation near the top of the butt. Gas production is demonstrated by the presence of bubbles or cracks in the medium.

Typical reactions observed on Kligler Iron Agar after 18-24 hours at 35°C.

Organism  Slope Butt Gas H2S
Shigella sonnei Red Yellow - -
Shigella dysenteriae Red Yellow - -
Salmonella typhi Red Yellow - +
Salmonella species Red Yellow + +
Enterobacter species Red Yellow + -
Klebsiella species Yellow Yellow + -
Escherichia coli Yellow Yellow + -
Proteus mirabilis Red Yellow - +
Morganella species Red Yellow V -
Citrobacter freundii Yellow Yellow + +
Yersinia species Red Yellow V -


Not all products are available for sale in all territories. Please inquire.
 
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Kligler Iron Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 9.1L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.