missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MacConkey Broth (Dehydrated)

Product Code. 10147812
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10147812 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10147812 Supplier Thermo Scientific™ Supplier No. CM0005B
Reliable Agri Testing Solutions
Use promo code 28024
View all promotions
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Identify coliforms from water and milk samples with this differential medium.

Identify coliforms from water and milk samples with differential Thermo Scientific™ Oxoid™ MacConkey Broth (Dehydrated). MacConkey Broth is the standard medium for the primary isolation of coliform bacteria, and has been recommended for this purpose by the Public Health Laboratory Service Water Committee1 and the World Health Organization.2 MacConkey Broth also conforms to the formulation specified by the UK Dept. of Health3 for milk grading.

Use MacConkey Broth for the cultivation of pathogens specifically. MacConkey Broth has been the standard medium for the primary isolation of coliform bacteria, which may be present in a variety of specimens.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description MacConkey Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 12.5L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.