missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Malt Extract

Product Code. 13108346
Change view
Click to view available options
Quantity:
500 g
5 kg
Unit Size:
500g
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13108346 5 kg 5kg
10188572 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13108346 Supplier Thermo Scientific™ Supplier No. LP0039T
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Oxoid Malt Extract is recommended for use in media for yeast and mold growth.

An extract of sprouted grain with conserved nitrogen and carbohydrate constituents
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Product Type Malt Extract
Quantity 5 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.