Learn More
Thermo Scientific™ Oxoid™ Meropenem Selective Supplement
Product Code. 10178492
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
Product Code. Quantity unitSize
10178492 10/Pk. Pack of 10
1 options
Product Code. 10178492
Brand: Thermo Scientific™ SR0184E
Used for the isolation of Vancomycin-Resistant Enterococci (VRE) and HighLevel Aminoglycoside-Resistant Enterococci (HLARE) from clinical specimens.
Oxoid Meropenem Selective Supplement (1mg/vial) is used for the isolation of Vancomycin-Resistant Enterococci (VRE) and HighLevel Aminoglycoside-Resistant Enterococci (HLARE) from clinical specimens.
  • Add to VRE Agar Base, Part No. CM0984B
  • Each vial supplements 500mL of medium
TRUSTED_SUSTAINABILITY
Specifications
Description Meropenem Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium