missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Metronidazole Disc

Product Code. 10200253
Change view
Click to view available options
Quantity:
50 Disks
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10200253 50 Disks Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10200253 Supplier Thermo Scientific™ Supplier No. DD0008T
  • ONLINE EXCLUSIVE PRICE
    9.40 €
    9.13 € / Pack of 50
    Save 0.27 €
    2% Off
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ Oxoid Metronidazole Disc is used for presumptive identification of Gardnerella vaginalis.

Thermo Scientific™ Oxoid Metronidazole Disc is used for presumptive identification of Gardnerella vaginalis.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Description Metronidazole Disk
Detectable Analytes G. vaginalis
Format Disk, Cartridge
Type Metronidazole Disk
Packaging Type 1 Cart./Pk.
Quantity 50 Disks
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.