missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Microbact™ GNB Reagents and Accessories

Product Code. 10036932
Change view
Click to view available options
Description:
Microbact Holding Tray 12A/12B
Microbact Mineral Oil
Microbact Oxidase Strip
Microbact Reagent Indole-Kovacs
Microbact Reagent Nitrate A
Microbact Reagent Nitrate B
Microbact Reagent Set D
Microbact Reagent Spot Indole DMACA
Microbact Reagent TDA
Microbact Reagent VP I
Microbact Reagent VP II
Quantity:
1 Bottle
1 Tray
10 mL/Bottle
50/Pk.
6 x 10 mL Bottles
Unit Size:
10mL
50 tests
50mL
Each
Pack of 10
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Quantity Unit Size
10036932 Microbact Reagent VP I 10 mL/Bottle 10mL
10726983 Microbact Reagent VP II 10 mL/Bottle 10mL
10331743 Microbact Oxidase Strip 50/Pk. 50 tests
12769030 Microbact Holding Tray 12A/12B 1 Tray Each
10371783 Microbact Reagent TDA 10 mL/Bottle 10mL
12749020 Microbact Reagent Set D 6 x 10 mL Bottles Each
10261063 Microbact Reagent Nitrate B 10 mL/Bottle 10mL
13206189 Microbact Reagent Spot Indole DMACA 10 mL/Bottle 10mL
13245769 Microbact Mineral Oil 1 Bottle 50mL
10600975 Microbact Reagent Indole-Kovacs 10 mL/Bottle 10mL
10158442 Microbact Reagent Nitrate A 10 mL/Bottle Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10036932 Supplier Thermo Scientific™ Supplier No. MB0181A
  • ONLINE EXCLUSIVE PRICE
    67.39 €
    63.50 € / 10mL
    Save 3.89 €
    5% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Simplify the detection of Gram-negative bacteria with convenient Thermo Scientific™ Oxoid™ Microbact™ GNB Reagents and Accessories.

Gram-negative bacteria are of significant concern in both clinical infections and food contamination. Identification of the causative agent is crucial in the clinical setting to identify the cause of infection and to establish an appropriate treatment regime. In the food industry, biochemical identification can help to trace the source of contamination. With these GNB reagents and accessories, Enterobacteriaceae and common miscellaneous Gram-negative bacilli are quickly and accurately identified so that proper, fast treatment for the patient may follow.

  • Fast: More results available overnight
  • Simple: Simple test strip or microplate format
  • Easy to Read and Interpret: Results are clearly visible as distinct color reactions that can be interpreted using the Microbact Identification Package (MB1244A)
  • Comprehensive: Identifies >100 Gram-negative bacteria, more unusual Enterobacteriaceae and oxidase-positive, Gram-negative bacteria
  • Flexible: Use single strips or combine systems for more complex identifications. Ideal for use in clinical or food microbiology laboratories
  • Reliable: A comprehensive range of biochemical tests based on published reference methodologies1-4

Additional items required: Mineral Oil (MB1093A), Oxidase Strips (MB0266A), Microbact Identification Package (Windows™) (MB1244A), Indole Reagent (MB0209A), VPI Reagent (MB0181A), VPII Reagent (MB0184A), TDA Reagent (MB0180A), NIT A Reagent (MB0186A), NIT B Reagent (MB0187A). The 6 reagents above are also available as Reagent Set D (MB1082A).

Not all products are available for sale in all territories. Please inquire.

Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Description Microbact Reagent VP I
Format Bottle
Packaging Type 1 Box
Quantity 10 mL/Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.