Learn More
Thermo Scientific™ Oxoid™ Microbact™ Software Program
Product Code. 10108452
Change view
Click to view available options
Quantity:
1 CD
Unit Size:
Each
Product Code. Quantity unitSize
10108452 1 CD Each
1 options
Product Code. 10108452 Supplier Thermo Scientific™ Supplier No. MB1244A
  • ONLINE EXCLUSIVE PRICE
    156.00 €
    151.61 € / Pack of 1
    Save 4.39 €
    2% Off
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
Easily read and interpret Microbact results with the Thermo Scientific™ Oxoid™ Microbact™ Software Program.
Easily read and interpret Microbact results with the Thermo Scientific™ Oxoid™ Microbact™ Software Program. Results are clearly visible as distinct color reactions, creating individualized microcodes for more precise reporting.
Easily read and interpret results from Microbact GNB, Listeria and Staphylococcus kits with the Microbact Software Program.
Requires: Windows™ 2000 V2.03.
Not all products are available for sale in all territories. Please inquire.
Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Description Microbact Software Program
Quantity 1 CD