missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Microbact™ Staph Fast Blue Reagent Set

Product Code. 13265599
Change view
Click to view available options
Quantity:
2 x 10mL Bottles
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13265599 2 x 10mL Bottles Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13265599 Supplier Thermo Scientific™ Supplier No. MB1588A
  • ONLINE EXCLUSIVE PRICE
    106.53 €
    98.94 € / Pack of 2
    Save 7.59 €
    7% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Simplify the identification of staphylococci, including staphylococcus aureus and coagulase-negative staphylococci, with the Thermo Scientific™ Oxoid™ Microbact™ Staph Fast Blue Reagent Set.

Simplify the identification of staphylococci, including staphylococcus aureus and coagulase-negative staphylococci (CNS), with the Thermo Scientific™ Oxoid™ Microbact™ Staph Fast Blue Reagent Set. This set is used in the Beta-galactosidase test for fast, reliable and easy-to-read results.

Used in the Beta-galactosidase test, this reagent set allows for quick, reliable and easy identification of staphylococci, including staphylococcus aureus and coagulase-negative staphylococci (CNS).

Not all products are available for sale in all territories. Please inquire.

Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Description Microbact Staph Fast Blue Reagent Set
Detectable Analytes Staphylococcus species
Type Reagent Set for Staphylococcus aureus Test Kit
Packaging Type 1 Box
Quantity 2 x 10mL Bottles
Target Organism Class Staphylococcus aureus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.