Learn More
Thermo Scientific™ Oxoid™ Mupirocin Antimicrobial Susceptibility discs
Product Code. 11983042
Change view
Click to view available options
Concentration:
200 μg
5 μg
Unit Size:
Pack of 250
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Concentration unitSize
11983042 200 μg Pack of 250
11933822 5 μg Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11983042 Supplier Thermo Scientific™ Supplier No. CT0523B
  • ONLINE EXCLUSIVE PRICE
    42.63 €
    35.18 € / Pack of 250
    Save 7.45 €
    17% Off
View all promotions
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
Determine the susceptibility of microorganisms using manual AST methods and a full range of high quality, easy-to-use antimicrobial susceptibility testing discs.
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Mupirocin Antimicrobial Susceptibility discs.
Ease of use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results, but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Mupirocin
Certifications/Compliance Complies with EUCAST guidelines
Concentration 200 μg
Description Oxoid Mupirocin Antimicrobial Susceptibility Discs
Antibiotic Code MUP
Product Type Antimicrobial Susceptibility Discs
Test Type Antimicrobial Susceptibility
No. per Pack 5 Cartridges/Pk.
Packaging Type 50 Discs/Cartridge
Quantity 50 Discs/Cartridge