missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Mycoplasma Supplement G

Product Code. 12751850
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12751850 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12751850 Supplier Thermo Scientific™ Supplier No. SR0059C
 Restricted Item
Estimated Shipment: 26-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Oxoid Mycoplasma Supplement is for the isolation of mycoplasmas.

Oxoid Mycoplasma Supplement G is for the isolation of mycoplasmas.

  • Add to Mycoplasma Pneumoniae Agar, Part No. CM0401B or CM0403B
  • Each vial supplements 80mL of medium
TRUSTED_SUSTAINABILITY

Specifications

Description Mycoplasma Supplement G
Form Freeze-dried
Product Type Microbiology Media
Quantity 10/Pk.
Yield Each vial for 80mL medium
Species Mycoplasma species
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.