missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Biocemical Identification System (O.B.I.S.) PYR

Product Code. 11483969
Change view
Click to view available options
Quantity:
60 Tests/Kit
Unit Size:
60 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483969 60 Tests/Kit 60 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11483969 Supplier Thermo Scientific™ Supplier No. ID0580M
  • ONLINE EXCLUSIVE PRICE
    183.92 €
    171.85 € / 60 tests
    Save 12.07 €
    6% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapidly determine PYRase activity is streptococci and Citrobacter spp. with this biochemical identification system.

Rapidly determine PYRase activity is streptococci and Citrobacter spp. with Thermo Scientific™ Oxoid™ Biochemical Identification System (O.B.I.S.) PYR. This rapid colorimetric test delivers simple and rapid results for the determination of PYRase activity in streptococci and Citrobacter spp, offering a rapid diagnostic alternative to time-consuming culture methods. In addition, the same enzymatic activity may be used to aid differentiation of Salmonella spp. from Citrobacter spp and other Enterobacteriaceae1,2.

Use O.B.I.S. PYR as a rapid diagnostic alternative to time-consuming culture methods.

  • Simple—easy to implement
  • Rapid—results in 6 minutes of testing suspect colonies
  • Non-carcinogenic substrates

The O.B.I.S. - PYR test uses Test Cards impregnated with L-pyroglutamic acid 7-amino-4-methyl-coumarin (7AMC) and dimethylaminocinnamaldehyde for the detection of PYRase activity. The enzymatic hydrolysis of this substrate by enterococci, Group A streptococci and Citrobacter spp. produces a purple color following the addition of the Developing Solution3.

We have developed a new system for PYRase testing4 using a non-carcinogenic substrate in response to health concerns associated with the use of β-naphthylamide (a potent carcinogen)5. The PYRase area on the O.B.I.S. PYR test is impregnated with L-pyroglutamic acid 7-amino-4-methylcoumarin (7AMC)4. Dimethylaminocinnamaldehyde is used as a color development reagent. The enzymatic hydrolysis of the substrate produces a purple color on addition of the O.B.I.S. PYR Developing Solution4.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description O.B.I.S. PYR
Detectable Analytes PYRase activity
Includes Test cards, developing solution, buffer solution
Quantity 60 Tests/Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.