missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ OGYE Selective Supplement

Product Code. 11424159
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424159 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11424159 Supplier Thermo Scientific™ Supplier No. SR0073A
  • ONLINE EXCLUSIVE PRICE
    58.40 €
    49.78 € / Pack of 10
    Save 8.62 €
    14% Off
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Oxoid OGYE Selective Supplement (Oxytetracycline Glucose Yeast Extract) is added to Oxytetracycline Glucose Yeast Extract Agar Base, Part No. CM0545B.

Oxoid OGYE Selective Supplement (Oxytetracycline Glucose Yeast Extract) is added to Oxytetracycline Glucose Yeast Extract Agar Base, Part No. CM0545B.

  • Add to Oxytetracycline Glucose Yeast Extract Agar Base, Part No. CM0545B
  • Each vial supplements 500mL of medium
TRUSTED_SUSTAINABILITY

Specifikationer

Description OGYE Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.