missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Peptone Bacteriological

Product Code. 10404013
Change view
Click to view available options
Quantity:
500 g
5 kg
Unit Size:
500g
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10404013 500 g 500g
10321923 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10404013 Supplier Thermo Scientific™ Supplier No. LP0037B
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Oxoid Peptone Bacteriological is an all-purpose peptone specially prepared for use in fermentation applications.

Thermo Scientific™ Oxoid Peptone Bacteriological and its neutralized form are very nutritious, all-purpose peptones prepared by the enzymatic digestion of selected animal protein sources. They are specially prepared to be compatible with other refined culture media or fermentation ingredients.

The combination of pancreatin and papain enzyme systems ensures that these bacteriological peptones contain a wide spectrum of polypeptides, reflected in their broad molecular profiles. 

The neutralized form evolved from the original to meet those occasions when a slightly higher pH is required. Both, when reconstituted, give a solution free of haze, cloudiness, or precipitation.

A nutritious, all-purpose peptone specially prepared for use with other refined culture media ingredients
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Dehydrated Powder, Ambient & Dry Storage
Form Dehydrated Powder
Product Type Peptone
For Use With (Application) Culture media or fermentation processes
Quantity 500 g
Protein Source Beef

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.