missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Purified Agar

Product Code. 10260343
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10260343 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10260343 Supplier Thermo Scientific™ Supplier No. LP0028B
  • ONLINE EXCLUSIVE PRICE
    316.00 €
    296.04 € / 500g
    Save 19.96 €
    6% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Oxoid Purified Agar is an exceptionally bright and clear agar recommended for electrophoresis studies but also suitable for many bacteriological purposes.

Oxoid Purified Agar is an exceptionally bright and clear agar recommended for electrophoresis studies but also suitable for many bacteriological purposes.

TRUSTED_SUSTAINABILITY

Specifications

Description Dehydrated; Purified agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 10.3L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.