Learn More
Thermo Scientific™ Oxoid™ Replacement Desiccant, Dispenser
Product Code. 10200343
Change view
Click to view available options
Product Type:
Replacement Desiccant
Unit Size:
Each
Product Code. Product Type Unit Size
10200343 Replacement Desiccant Each
1 options
Product Code. 10200343 Supplier Thermo Scientific™ Supplier No. ST9100
  • ONLINE EXCLUSIVE PRICE
    49.40 €
    46.01 € / Pack of 1
    Save 3.39 €
    6% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Maintain a low moisture environment when storing AST products used for manual Antimicrobial Susceptibility Testing.
Maintain a low moisture environment when storing AST products for manual Antimicrobial Susceptibility Testing with Thermo Scientific™ Oxoid™ Replacement Desiccant (Dispenser).
The Oxoid Replacement Desiccant fits both the Oxoid 8- and 12-position antimicrobial susceptibility disc dispensers.
One of the simplest and most widely used methods for susceptibility testing, disc diffusion is extremely flexible, offering a wide variety of antibiotics and concentrations which can easily be changed as required.
Disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Description Oxoid Replacement Desiccant (Dispenser)
Product Type Replacement Desiccant