missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ RPF Supplement

Product Code. 10681515
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10681515 10/Pk. 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10681515 Supplier Thermo Scientific™ Supplier No. SR0122A
  • ONLINE EXCLUSIVE PRICE
    357.00 €
    347.21 € / 1000mL
    Save 9.79 €
    2% Off
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The Oxoid RPF Supplement (Rabbit Plasma Fibrinogen) is used for isolation and enumeration of coagulase positive staphylococci from food.

Oxoid RPF Supplement (Rabbit Plasma Fibrinogen) is used for the isolation and enumeration of coagulase positive staphylococci from food.

  • Use only with Baird-Parker Agar Base, RPF, Part No. CM0961B
  • Each vial supplements 100mL of medium
TRUSTED_SUSTAINABILITY

Specifications

Description RPF Supplement
Form Freeze-dried
Product Type Microbiology Media
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.