missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Saline Tablets
Click to view available options
Quantity:
Each
Unit Size:
100 tablets
Description
Oxoid Saline Tablets is used for preparation of 500mL of 0.85% isotonic saline.
Specifications
Specifications
| Description | Saline Tablets |
| Form | Tablet |
| Product Type | Media Supplement |
| Quantity | Each |
| Yield | Each tab for 500mL medium |
For Laboratory Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction