missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Sensitest Agar

Product Code. 11963952
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
Product Code. Quantity unitSize
11963952 500 g 500g
1 options
This item is not returnable. View return policy

Product Code. 11963952

Brand: Thermo Scientific™ CM0409B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Oxoid Sensitest Agar is used for antimicrobial susceptibility testing.

Oxoid Sensitest Agar is used for antimicrobial susceptibility testing.

TRUSTED_SUSTAINABILITY

Specifications

Description Sensitest Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 15.6L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.