Learn More
Thermo Scientific™ Oxoid™ SPS Disc
Product Code. 10127462
Change view
Click to view available options
Quantity:
50 Disks/Cartridge
Unit Size:
Pack of 50
Product Code. Quantity Unit Size
10127462 50 Disks/Cartridge Pack of 50
1 options
Product Code. 10127462 Supplier Thermo Scientific™ Supplier No. DD0016T
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Thermo Scientific™ Oxoid SPS Disc is used for presumptive identification of Peptostreptococcus anaerobius.
Thermo Scientific™ Oxoid SPS Disc is used for presumptive identification of Peptostreptococcus anaerobius.
Susceptibility disc format
TRUSTED_SUSTAINABILITY
Specifications
Description SPS Disk
Detectable Analytes P. anaerobius
Format Susceptibility Disk
Type SPS Disk
Quantity 50 Disks/Cartridge