missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Staph/Strep Selective Supplement
Description
Oxoid Staph/Strep Selective Supplement is used for preparation of Columbia CNA Agar.
- Add to Columbia Blood Agar Base Part No. CM0331
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | Staph/Strep Selective Supplement |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 500 g |
| Yield | Each vial for 500mL medium |
| Species | Staphylococcus species, Streptococcus species |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?