missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Sulphonamide Disc

Product Code. 10629574
Change view
Click to view available options
Quantity:
50 Disks/Cartridge
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10629574 50 Disks/Cartridge Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10629574 Supplier Thermo Scientific™ Supplier No. DD0011T
  • ONLINE EXCLUSIVE PRICE
    9.90 €
    9.63 € / Pack of 50
    Save 0.27 €
    2% Off
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Thermo Scientific™ Oxoid Sulphonamide Disc is used for identification of Gardnerella vaginalis.

Thermo Scientific™ Oxoid Sulphonamide Disc is used for identification of Gardnerella vaginalis.

  • 100μg
  • 50 Discs/Cartridge
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Sulphonamide Disk
Detectable Analytes G. vaginalis
Format Disk, Cartridge
Type Sulphonamide Disk
Packaging Type 1 Cart./Pk.
Quantity 50 Disks/Cartridge
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.