missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ TST-RPLA Kit

Product Code. 11488299
Change view
Click to view available options
Quantity:
20 Tests/Kit
Unit Size:
20 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488299 20 Tests/Kit 20 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11488299 Supplier Thermo Scientific™ Supplier No. TD0940A

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Oxoid TST-RPLA Kit detects staphylococcal toxic-shock syndrome toxin (TST) by reversed passive latex agglutination.

Oxoid TST-RPLA Kit detects staphylococcal toxic-shock syndrome toxin (TST) by reversed passive latex agglutination.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Product Type Microbiology Media
Test Type Reverse Passive Latex Agglutination
Quantity 20 Tests/Kit
Specimen Type Staphylococcal Toxic-Shock Syndrome Toxin (TST)
Species Staphylococcus species
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.