missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Vegetable Peptone Broth

Product Code. 10066842
Click to view available options
Quantity:
500 g
5 kg
Unit Size:
500g
5kg
This item is not returnable. View return policy

Product Code. 10066842

Brand: Thermo Scientific™ VG0101B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ Oxoid Vegetable Peptone Broth is a meat-free general-purpose peptone.

Thermo Scientific™ Oxoid Vegetable Peptone Broth is a highly nutritious, general-purpose peptone.

An exceptional meat-free alternative to Typtone Soya Broth, it delivers equivalent performance in culture media or as a fermentation base.

  • Meat-free alternative to Typtone Soya Broth
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Dehydrated Powder, Ambient & Dry Storage
Form Dehydrated Powder
Product Type Peptone
For Use With (Application) Culture media or fermentation processes
Quantity 500 g
Protein Source Pea / Yeast

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.