missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Vogel-Johnson Agar (Dehydrated)

Product Code. 10527863
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10527863 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10527863 Supplier Thermo Scientific™ Supplier No. CM0641B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate and presumptively identify Staphylococcus aureus from clinical specimens and food with this selective medium.

Isolate and presumptively identify Staphylococcus aureus from clinical specimens and food with Thermo Scientific™ Oxoid™ Vogel-Johnson Agar (Dehydrated). The medium is based on the formula corresponding to the United States Pharmacopoeia1. Staphylococcus aureus reduces tellurite to metallic tellurium in the medium and appear as black colonies.

Staphylococcus aureus is a Gram-positive, round-shaped bacteria and can cause skin and soft tissue infections such as abscesses (boils), furuncles, and cellulitis. Although most staph infections are not serious, S. aureus can cause serious infections such as bloodstream infections, pneumonia, or bone and joint infections.

Vogel-Johnson Agar allows the early detection and of Staphylococcus aureus from heavily contaminated foods and clinical specimens by selecting and identifying coagulase-positive and monitol-fermenting strains.

  • Selective: tellurite, lithium chloride and the high glycine concentration inhibits most of the contaminating organisms.
  • Easy-to-read: Staphylococcus aureus appear as black colonies due to their ability to reduce tellurite to metallic tellurium.
  • Conforms to the United States Pharmacopoeia1.
  • Allows the early detection and of Staphylococcus aureus from heavily contaminated foods and clinical specimens by selecting and identifying coagulase-positive and monitol-fermenting strains.

Vogel and Johnson2 modified the Tellurite Glycine Agar formula of Zebovitz et al.3 by doubling the mannitol concentration to 1% (w/v) and adding Phenol Red as a pH indicator. The enhanced fermentation reaction which occurs as a result of the increase in mannitol content is clearly indicated by the development of yellow zones surrounding the colonies.

Prolonged incubation may result in the growth of black coagulase-negative colonies and if these organisms also ferment mannitol they may be falsely identified from their appearance as Staphylococcus aureus. In these circumstances, further tests of identity should be carried out before concluding that the organism is Staphylococcus aureus.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: FDA.
Description Vogel-Johnson Agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 8.2L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.