missing translation for 'onlineSavingsMsg'
Learn More

Oxone∣r, monopersulfate

Product Code. 11463850
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
1 kg
5 kg
missing translation for 'unitSize'
100g
1kg
5kg
This item is not returnable. View return policy

Product Code. 11463850

missing translation for 'mfr': Thermo Scientific Alfa Aesar 089892.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 70693-62-8 | H3K5O18S4 | 614.74 g/mol

Oxone is used for halogenation of a,b-unsaturated carbonyl compounds and catalytic generation of hypervalent iodine reagents for alcohol oxidation. It is a used for rapid, and good synthesis of oxaziridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70693-62-8
Molecular Formula H3K5O18S4
Molecular Weight (g/mol) 614.74
MDL Number MFCD00040551
InChI Key HJKYXKSLRZKNSI-UHFFFAOYSA-I
Synonym Potassium peroxymonosulfate, Potassium monopersulfate triple salt

Specifications

Physical Form Granules
Quantity 1 kg
Solubility Information Soluble in water (100mg/ml).
Formula Weight 615.5
Sensitivity Hygroscopic
Density 1.15 g/mL
Chemical Name or Material Oxone
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Oxone, monopersulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 A
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Potassium peroxymonosulfate sulfate, Potassium pyrosulfate,Dipotassium peroxodisulphate, Potassium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.