missing translation for 'onlineSavingsMsg'
Learn More

Oxyberberin, TRC

Product Code. 30419127
Change view
Click to view available options
Quantity:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30419127 50 mg
30423035 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30419127 Supplier Toronto Research Chemicals Supplier No. TRCO87066550MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 549-21-3
Molecular Formula C20 H17 N O5
Molecular Weight (g/mol) 351.35
Synonym 8H-Benzo[g]-1,3-benzodioxolo[5,6-a]quinolizin-8-one, 5,6-dihydro-9,10-dimethoxy-, Berbin-8-one, 13,13a-didehydro-9,10-dimethoxy-2,3-(methylenedioxy)- (8CI), Oxyberberine (6CI), 8-Oxoberberine, 8-Oxyberberine, Berberin-8-one, Berlambine, JKL 1073A, NSC 93138
SMILES O=C1C=2C(OC)=C(OC)C=CC2C=C3C4=CC=5OCOC5C=C4CCN13

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C20H17NO5/c1-23-15-4-3-12-7-14-13-9-17-16(25-10-26-17)8-11(13)5-6-21(14)20(22)18(12)19(15)24-2/h3-4,7-9H,5-6,10H2,1-2H3
Formula Weight 351.1107
Chemical Name or Material Berlambine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.