missing translation for 'onlineSavingsMsg'
Learn More

Oxychlordane, TRC

Product Code. 30041074
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041074 10 mg 10mg
30040725 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30041074 Supplier Toronto Research Chemicals Supplier No. TRCO87057510MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27304-13-8
Molecular Formula C10 H4 Cl8 O
Molecular Weight (g/mol) 423.76
Synonym 2,5-Methano-2H-indeno[1,2-b]oxirene, 2,3,4,5,6,6a,7,7-octachloro-1a,1b,5,5a,6,6a-hexahydro-, (1aα,1bβ,2α,5α,5aβ,6β,6aα)-, 4,7-Methanoindan, 1,2,4,5,6,7,8,8-octachloro-2,3-epoxy-3a,4,7,7a-tetrahydro-, exo,endo- (8CI), rel-(1aR,1bS,2R,5S,5aR,6S,6aS)-2,3,4,5,6,6a,7,7-Octachloro-1a,1b,5,5a,6,6a-hexahydro-2,5-methano-2H-indeno[1,2-b]oxirene, (±)-Oxychlordane, Octachlor epoxide, Oxychlordan, Oxychlordane
SMILES Cl[C@@H]1[C@@H]2[C@H]([C@@H]3O[C@]13Cl)[C@]4(Cl)C(=C(Cl)[C@@]2(Cl)C4(Cl)Cl)Cl

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H4Cl8O/c11-3-1-2(6-9(3,16)19-6)8(15)5(13)4(12)7(1,14)10(8,17)18/h1-3,6H/t1-,2+,3+,6-,7+,8-,9+/m0/s1
Formula Weight 419.777
Percent Purity >95%
Chemical Name or Material oxy-Chlordane
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.