Learn More
WTW™ Oxygen Test Kits
Product Code. 11774745
Change view
Click to view available options
Range:
1 at 20°C
Unit Size:
Pack of 25
Product Code. Range Unit Size
11774745 1 at 20°C Pack of 25
1 options
Product Code. 11774745 Supplier WTW™ Supplier No. 250403
  • ONLINE EXCLUSIVE PRICE
    201.00 €
    199.00 € / Pack of 25
    Save 2.00 €
    1.00% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
For Use With (Application) Surface water, drinking water
For Use With (Equipment) PhotoLab™6, PhotoLab™ S12, PhotoLab™ 6000, PhotoLab™ Spektral photometer
No. of Tests 25
Range 1 at 20°C
Type COD Chemical oxygen demand O2 cell test kit