missing translation for 'onlineSavingsMsg'
Learn More

Oxymetazoline Hydrochloride, TRC

Product Code. 30416803
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416803 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30416803 Supplier Toronto Research Chemicals Supplier No. TRCO876470100MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxymetazoline Hydrochloride
CAS 2315-02-8
Molecular Formula C16 H24 N2 O . Cl H
Quantity 100 mg
Synonym Oxymetazoline Hydrochloride, 3-[(4,5-Dihydro-1H-imidazol-2-yl)methyl]-6-(1,1-dimethylethyl)-2,4-dimethylphenol hydrochloride (1:1), 3-[(4,5-Dihydro-1H-imidazol-2-yl)methyl]-6-(1,1-dimethylethyl)-2,4-dimethylphenol monohydrochloride, 6-tert-Butyl-3-(2-imidazolin-2-ylmethyl)-2,4-dimethylphenol hydrochloride, 6-tert-Butyl-3-(2-imidazolin-2-ylmethyl)-2,4-dimethylphenol monohydrochloride, 2,6-Dimethyl-4-tertiarybutyl-3-hydroxyphenyl)methylimidazoline hydrochloride, 2-(4-tert-Butyl-2,6-dimethyl-3-hydroxybenzyl)-2-imidazolinium chloride, 4-Way Nasal 12 Hour Spray, 4-Way Nasal Spray, 6-tert-Butyl-3-(2-imidazolin-2-ylmethyl)-2,4-dimethylphenol hydrochloride, Afrazine, Afrin, Afrin hydrochloride, Allerest 12 Hour Nasal Spray, Dristan Long-Lasting Nasal Mist, Duration, Duration 12 Hour Nasal Spray, H 990, Iliadin, Nafrine, Nasivin, Neo-Synephrine 12 Hour, Neo-Synephrine 12 Hour NTZ, Nostrilla, Ocuclear, Oxilin, Oxymetazoline chloride, Oxymetazoline hydrochloride, Respir, Sch 9384
InChI Formula InChI=1S/C16H24N2O.ClH/c1-10-8-13(16(3,4)5)15(19)11(2)12(10)9-14-17-6-7-18-14;/h8,19H,6-7,9H2,1-5H3,(H,17,18);1H
SMILES Cl.Cc1cc(c(O)c(C)c1CC2=NCCN2)C(C)(C)C
IUPAC Name 6-tert-butyl-3-(4,5-dihydro-1H-imidazol-2-ylmethyl)-2,4-dimethylphenol;hydrochloride
Molecular Weight (g/mol) 296.84
Formula Weight 296.1655
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.