missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Oxytocin acetate

Product Code. 16411276
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
Unit Size:
200mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16411276 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16411276 Supplier MedChemExpress Supplier No. HY17571A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Oxytocin acetate is a pleiotropic, hypothalamic peptide known for facilitating parturition, lactation, and prosocial behaviors. Oxytocin acetate can function as a stress-coping molecule with anti-inflammatory, antioxidant, and protective effects especially in the face of adversity or trauma.

Specifications

Purity 99.0%
CAS 6233-83-6
Content And Storage Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Molecular Weight (g/mol) 1067.24 g/mol
Product Type Peptides
Quantity 200 mg
Shipping Condition Room temperature
Solubility H2O : 50 mg/mL (46.85 mM∣Need ultrasonic) ∣DMSO : ≥ 44 mg/mL (41.23 mM)
Synonym α-Hypophamine acetate∣ Oxytocic hormone acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.