missing translation for 'onlineSavingsMsg'
Learn More

p-Acetoxyacetophenone, TRC

Product Code. 30320593
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30320593 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320593 Supplier Toronto Research Chemicals Supplier No. TRCA0121005G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material p-Acetoxyacetophenone
CAS 13031-43-1
Molecular Formula C10H10O3
Quantity 5 g
Synonym 1-[4-(acetyloxy)phenyl]-Ethanone, 4’-hydroxy-Acetophenone acetate, 1-[4-(Acetyloxy)phenyl]ethanone, 4-Acetoxyacetophenone, 4-Acetylphenyl acetate, 4’-Acetoxyacetophenone, NSC 41974, p-Acetylphenyl acetate
InChI Formula InChI=1S/C10H10O3/c1-7(11)9-3-5-10(6-4-9)13-8(2)12/h3-6H,1-2H3
SMILES CC(=O)Oc1ccc(cc1)C(=O)C
IUPAC Name (4-acetylphenyl) acetate
Molecular Weight (g/mol) 178.18
Formula Weight 178.063
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.