missing translation for 'onlineSavingsMsg'
Learn More

p-Cymene, 97%

Product Code. 11499496
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499496 500 mL 500mL
11409506 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499496 Supplier Thermo Scientific Alfa Aesar Supplier No. A19226.AP
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 99-87-6 | C10H14 | 134.222 g/mol

It is used in heat-exchange media, metal polishes, and thinners for lacquers/varnishes. Also used to make para-cresol, carvacrol, and synthetic resins. It is also used as a component of commercial terpene solvent mixtures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in heat-exchange media, metal polishes, and thinners for lacquers/varnishes. Also used to make para-cresol, carvacrol, and synthetic resins. It is also used as a component of commercial terpene solvent mixtures.

Solubility
Soluble in water at 0.023g/L.20°C.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-87-6
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00008893
InChI Key HFPZCAJZSCWRBC-UHFFFAOYSA-N
Synonym p-cymene, 4-isopropyltoluene, p-isopropyltoluene, dolcymene, p-cymol, paracymene, cymene, para-cymene, 1-isopropyl-4-methylbenzene, camphogen
PubChem CID 7463
ChEBI CHEBI:28768
IUPAC Name 1-methyl-4-propan-2-ylbenzene
SMILES CC1=CC=C(C=C1)C(C)C

Specifications

Melting Point -68°C
Density 0.86
Boiling Point 176°C to 178°C
Flash Point 47°C (116°F)
Odor Aromatic
Refractive Index 1.49
Quantity 500 mL
UN Number UN2046
Beilstein 1903377
Merck Index 14,2763
Solubility Information Soluble in water at 0.023g/L.20°C.
Formula Weight 134.22
Percent Purity 97%
Chemical Name or Material p-Cymene
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • p-Cymene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H331-Toxic if inhaled.
  • H361-Suspected of damaging fertility or the unborn child.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: p-Cymene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.