missing translation for 'onlineSavingsMsg'
Learn More

p-Cymene, TRC

Product Code. 30003911
Change view
Click to view available options
Quantity:
5 mL
25 mL
50 mL
Unit Size:
25mL
50mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003911 25 mL 25mL
30009010 5 mL 5mL
30008031 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003911 Supplier Toronto Research Chemicals Supplier No. TRCC99410525ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-87-6
Molecular Formula C10 H14
Molecular Weight (g/mol) 134.22
Synonym p-Cymene (8CI), 1-Methyl-4-(1-methylethyl)benzene, 1-Isopropyl-4-methylbenzene, 1-Methyl-4-isopropylbenzene, 2-p-Tolylpropane, 4-Cymene, 4-Isopropyl-1-methylbenzene, 4-Isopropyltoluene, 4-Methylisopropylbenzene, Camphogen, Dolcymene, NSC 4162, p-Cymol, p-Isopropyltoluene, p-Methylcumene, p-Methylisopropylbenzene, para-Cymene
IUPAC Name 1-methyl-4-propan-2-ylbenzene
SMILES CC(C)c1ccc(C)cc1

Specifications

Quantity 25 mL
InChI Formula InChI=1S/C10H14/c1-8(2)10-6-4-9(3)5-7-10/h4-8H,1-3H3
Formula Weight 134.1096
Percent Purity >95%
Chemical Name or Material 4-Cymene
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.