missing translation for 'onlineSavingsMsg'
Learn More

p-Hydroxycinnamic acid, MedChemExpress

Product Code. 16472396
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16472396 500 mg 500mg
16439186 100 mg 100mg
16434947 50 mg 50mg
16419298 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16472396 Supplier Medchem Express Supplier No. HYN2391

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress p-Hydroxycinnamic acid, a common dietary phenol, could inhibit platelet activity, with IC50s of 371 μM, 126 μM for thromboxane B2 production and lipopolysaccharide-induced prostaglandin E2 generation, respectively.

p-Hydroxycinnamic acid, a common dietary phenol, could inhibit platelet activity, with IC50s of 371 μM, 126 μM for thromboxane B2 production and lipopolysaccharide-induced prostaglandin E2 generation, respectively.

Specifikationer

Chemical Name or Material p-Hydroxycinnamic acid
CAS 7400-08-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C9H8O3
Quantity 500 mg
Solubility Information DMSO : ≥ 100 mg/mL (609.16 mM) ∣Ethanol : 33.33 mg/mL (203.03 mM; Need ultrasonic)
SMILES O=C(O)/C=C/C1=CC=C(O)C=C1
Molecular Weight (g/mol) 164.16
Formula Weight 164.16
Percent Purity 98.24%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Visa mer Visa mindre

Research purposes only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.