missing translation for 'onlineSavingsMsg'
Learn More

p-Nitrophenyl Alpha-D-Mannopyranoside, TRC

Product Code. 30483942
Change view
Click to view available options
Quantity:
250 mg
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30483942 250 mg
30481116 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30483942 Supplier Toronto Research Chemicals Supplier No. TRCN503995250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10357-27-4
Molecular Formula C12 H15 N O8
Molecular Weight (g/mol) 301.25
Synonym α-D-Mannopyranoside, 4-nitrophenyl, Mannopyranoside, p-nitrophenyl (7CI), Mannopyranoside, p-nitrophenyl, α-D- (8CI), 4-Nitrophenyl α-D-mannopyranoside, 4'-Nitrophenyl α-D-mannopyranoside, p-Nitrophenyl α-D-mannopyranoside, p-Nitrophenyl α-mannoside
IUPAC Name (2R,3S,4S,5S,6R)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@H](Oc2ccc(cc2)[N+](=O)[O-])[C@@H](O)[C@@H](O)[C@@H]1O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H15NO8/c14-5-8-9(15)10(16)11(17)12(21-8)20-7-3-1-6(2-4-7)13(18)19/h1-4,8-12,14-17H,5H2/t8-,9-,10+,11+,12+/m1/s1
Formula Weight 301.0798
Chemical Name or Material 4-Nitrophenyl-alpha-D-mannopyranoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.