missing translation for 'onlineSavingsMsg'
Learn More

p-Nitrophenyl Beta-D-Xylopyranoside, TRC

Product Code. 30476444
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30476444 250 mg
30474525 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30476444 Supplier Toronto Research Chemicals Supplier No. TRCN509050250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2001-96-9
Molecular Formula C11 H13 N O7
Molecular Weight (g/mol) 271.22
IUPAC Name (2S,3R,4S,5R)-2-(4-nitrophenoxy)oxane-3,4,5-triol
SMILES O[C@@H]1CO[C@@H](Oc2ccc(cc2)[N+](=O)[O-])[C@H](O)[C@H]1O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C11H13NO7/c13-8-5-18-11(10(15)9(8)14)19-7-3-1-6(2-4-7)12(16)17/h1-4,8-11,13-15H,5H2/t8-,9+,10-,11+/m1/s1
Formula Weight 271.0692
Chemical Name or Material 4-Nitrophenyl-beta-D-xylopyranoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.