missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford P-Nitrophenyl Phosphate, bis-CHA Salt
Click to view available options
Quantity:
1 g
Unit Size:
1g
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Free PNP: <250ppm
Specifications
Specifications
| Quantity | 1 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction