missing translation for 'onlineSavingsMsg'
Learn More

p-Nitrophenyl Phosphate (Disodium Salt, Hexahydrate), Fisher BioReagents

Product Code. 10697525
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
50 g
Packaging:
Amber Glass
Unit Size:
10g
1g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10697525 10 g Amber Glass 10g
10542965 1 g Amber Glass 1g
10246294 5 g Amber Glass 5g
10656765 50 g Amber Glass 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10697525 Supplier Fisher BioReagents Supplier No. BP253410
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
p-Nitrophenyl Phosphate is suitable for use as a substrate for alkaline and acid phosphatase.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4264-83-9
Molecular Formula C6H4NNa2O6P
Molecular Weight (g/mol) 263.05
MDL Number MFCD00007319
InChI Key VIYFPAMJCJLZKD-UHFFFAOYSA-L
Synonym pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system
PubChem CID 77949
IUPAC Name disodium;(4-nitrophenyl) phosphate
SMILES [Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1

Specifications

Melting Point 300°C
Color White
Assay Percent Range ≥98.0%
Packaging Amber Glass
Quantity 10 g
Formula Weight 371
Physical Form Powder or Solid
Chemical Name or Material p-Nitrophenyl Phosphate Disodium Salt, Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.