missing translation for 'onlineSavingsMsg'
Learn More

p-Phenylene diisothiocyanate, 99%

Product Code. 10701072 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10701072 25 g Glass bottle 25g
10177560 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10701072 Supplier Thermo Scientific Acros Supplier No. 417510250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4044-65-9 | C8H4N2S2 | 192.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4044-65-9
Molecular Formula C8H4N2S2
Molecular Weight (g/mol) 192.27
MDL Number MFCD00004811
InChI Key OMWQUXGVXQELIX-UHFFFAOYSA-N
Synonym bitoscanate, 1,4-phenylene diisothiocyanate, jonit, p-phenylene diisothiocyanate, benzene, 1,4-diisothiocyanato, bitoscanat, bitoscanate inn, phenylene thiocyanate, bitoscanatum latin, 1,4-phenylenediisothiocyanate
PubChem CID 19958
IUPAC Name 1,4-diisothiocyanatobenzene
SMILES C1=CC(=CC=C1N=C=S)N=C=S

Specifications

Melting Point 129.0°C to 133.0°C
Color Beige-Gray to White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula SCNC6H4NCS
Quantity 25 g
Merck Index 15, 1309
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and chloroform,slightly soluble in acetone
Formula Weight 192.27
Percent Purity 99%
Physical Form Crystalline Needles or Powder
Chemical Name or Material p-Phenylene diisothiocyanate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • p-Phenylene diisothiocyanate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.