missing translation for 'onlineSavingsMsg'
Learn More

p-Phenylenediamine, 97%

Product Code. 11447627
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11447627 5000 g 5000g
11437627 250 g 250g
11427627 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11447627 Supplier Thermo Scientific Alfa Aesar Supplier No. A15680.0I
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 106-50-3 | C6H8N2 | 108.144 g/mol

p-Phenylenediamine is used as a precursor to aramid plastics, Kevlar fibers and to urethane polymers. It serves as a developing agent in photography, antioxidants in rubber products and as a vulcanization accelerator. It is used as a henna surrogate for temporary tattoos and as a histological stain for lipids such as myelin. It is an anti-fade reagent used in fluorescence microscopy to retard photobleaching of fluorescein and other fluorescent dyes. Furthermore, it is also an ingredient in hair dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Phenylenediamine is used as a precursor to aramid plastics, Kevlar fibers and to urethane polymers. It serves as a developing agent in photography, antioxidants in rubber products and as a vulcanization accelerator. It is used as a henna surrogate for temporary tattoos and as a histological stain for lipids such as myelin. It is an anti-fade reagent used in fluorescence microscopy to retard photobleaching of fluorescein and other fluorescent dyes. Furthermore, it is also an ingredient in hair dyes.

Solubility
Soluble in alcohol, chloroform, ether and hot benzene.

Notes
Store in cool place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-50-3
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00007901
InChI Key CBCKQZAAMUWICA-UHFFFAOYSA-N
Synonym p-phenylenediamine, 1,4-benzenediamine, 1,4-diaminobenzene, 1,4-phenylenediamine, 4-aminoaniline, para-phenylenediamine, p-aminoaniline, p-diaminobenzene, orsin, 4-phenylenediamine
PubChem CID 7814
ChEBI CHEBI:51403
IUPAC Name benzene-1,4-diamine
SMILES C1=CC(=CC=C1N)N

Specifications

Melting Point 138°C to 142°C
Boiling Point 267°C to 271°C
Flash Point 156°C (312°F)
Assay Percent Range 97%
Odor Weak
Quantity 5000 g
UN Number UN1673
Beilstein 742029
Merck Index 14,7285
Solubility Information Soluble in alcohol,chloroform,ether and hot benzene.
Formula Weight 108.14
Percent Purity 97%
Chemical Name or Material p-Phenylenediamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.