missing translation for 'onlineSavingsMsg'
Learn More

p-Phenylenediamine dihydrochloride, 99+%

Product Code. 10561724 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10561724 25 g Glass bottle 25g
10045870 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10561724 Supplier Thermo Scientific Acros Supplier No. 417480250
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 624-18-0 | C6H8N2·2HCl | 181.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-18-0
Molecular Formula C6H8N2·2HCl
Molecular Weight (g/mol) 181.06
MDL Number MFCD00013002
InChI Key IYXMNTLBLQNMLM-UHFFFAOYSA-N
Synonym benzene-1,4-diamine dihydrochloride, p-phenylenediamine dihydrochloride, 1,4-diaminobenzene dihydrochloride, 1,4-phenylenediamine dihydrochloride, fourrine ds, 1,4-benzenediamine, dihydrochloride, pelagol cd, pelagol grey cd, durafur black rc, fourrine 64
PubChem CID 12205
IUPAC Name benzene-1,4-diamine;dihydrochloride
SMILES C1=CC(=CC=C1N)N.Cl.Cl

Specifications

Color Gray-Purple to White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (Argentometry)
Packaging Glass bottle
Linear Formula C6H4(NH2)2·2HCl
Quantity 25 g
Solubility Information Solubility in water: soluble.
Formula Weight 181.06
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material p-Phenylenediamine dihydrochloride, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • p-Phenylenediamine dihydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.