missing translation for 'onlineSavingsMsg'
Learn More

p-Phenylenediamine sulfate, 98%, Thermo Scientific™

Product Code. 11470523
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470523 5000 g 5000g
11460523 250 g 250g
11450523 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470523 Supplier Thermo Scientific Alfa Aesar Supplier No. A12659.0I
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16245-77-5
Molecular Formula C6H10N2O4S
Molecular Weight (g/mol) 206.22
MDL Number MFCD00035510
InChI Key UFPKLWVNKAMAPE-UHFFFAOYSA-N
Synonym p-phenylenediamine sulfate, benzene-1,4-diamine sulfate, 1,4-diaminobenzene sulfate, 1,4-phenylenediamine sulfate, 1,4-benzenediamine, sulfate 1:1, unii-s7igm30961, benzene-1,4-diamine; sulfuric acid, 1,4-benzenediamine, sulfate, benzene-1,4-diammonium sulphate, 1,4-benzenediamine sulfate
PubChem CID 27769
IUPAC Name benzene-1,4-diamine;sulfuric acid
SMILES OS(O)(=O)=O.NC1=CC=C(N)C=C1

Specifications

Melting Point >300°C
Quantity 5000 g
UN Number UN2811
Sensitivity Hygroscopic
Formula Weight 206.22
Percent Purity 98%
Chemical Name or Material p-Phenylenediamine sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.